N-Methyl-3-nitroaniline 98%

Supplier: Ambeed
Warning

A489899-100MG A489899-1G A489899-250MG
76830-058EA 25.47 USD
76830-058 76830-060 76830-062
N-Methyl-3-nitroaniline 98%
N-Methyl-3-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.153 g/mol
MDL Number: MFCD00963641
CAS Number: 619-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR