2-Chloroterephthalic acid 98%

Supplier: Ambeed
Warning

A489536-100MG A489536-1G A489536-5G
76830-050EA 18 USD
76830-050 77551-480 77886-268
2-Chloroterephthalic acid 98%
2-Chloroterephthalic acid

Precaución: for Research Use Only

Formula: C₈H₅ClO₄
MDL Number: MFCD14635792
CAS Number: 1967-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR