4-Bromo-6-fluoroquinoline 98%

Supplier: Ambeed
Danger

A489314-1G A489314-250MG A489314-5G
76830-044EA 34 USD
76830-044 76830-046 76830-048
4-Bromo-6-fluoroquinoline 98%
4-Bromo-6-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MDL Number: MFCD09261094
CAS Number: 661463-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR