Potassium (1-naphthalene)trifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(naphthalen-1-yl)borate, (1-Naphthalene)trifluoroboric acid potassium salt

A489116-25G A489116-5G
76830-040EA 142 USD
76830-040 76830-042
Potassium (1-naphthalene)trifluoroborate 98%
Potassium (1-naphthalene)trifluoroborate

Precaución: for Research Use Only

Formula: C₁₀H₇BF₃K
MW: 234.07
MDL Number: MFCD06659936
CAS Number: 166328-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR