3,3-Diphenylacrylic acid 95%

Supplier: Ambeed
Warning

A488817-100MG A488817-250MG A488817-1G
76830-030EA 59 USD
76830-030 76830-032 77886-267
3,3-Diphenylacrylic acid 95%
3,3-Diphenylacrylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₂
MDL Number: MFCD00137211
CAS Number: 606-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR