4-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Trifluoromethyl)picolinimidamide hydrochloride
A485809-100MG
A485809-1G
A485809-250MG
76830-018EA
128.37
USD
76830-018
76830-020
76830-022
4-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride 95%
4-(Trifluoromethyl)pyridine-2-carboxamidine hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₆F₃N₃·ClH MW: 225.599 g/mol |
MDL Number:
MFCD12755592 CAS Number: 909109-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...