5-Phenyl-1,3-oxazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A483398-100MG A483398-250MG A483398-5G
76830-008EA 11 USD
76830-008 76830-010 76830-012
5-Phenyl-1,3-oxazole-4-carboxylic acid 97%
5-Phenyl-1,3-oxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₃
MDL Number: MFCD00105447
CAS Number: 99924-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR