tert-Butyl 3-methylpyrazole-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-methyl-1H-pyrazole-1-carboxylate

A478728-1G A478728-250MG A478728-5G
76829-978EA 66.84 USD
76829-978 76829-980 76829-982
tert-Butyl 3-methylpyrazole-1-carboxylate 95%
tert-Butyl 3-methylpyrazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
MDL Number: MFCD21609566
CAS Number: 186551-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR