N-(Phenylsulfonyl)glycine Methyl Ester 98%

Supplier: Ambeed
Warning

A478525-100MG A478525-1G A478525-250MG
76829-972EA 13 USD
76829-972 76829-974 76829-976
N-(Phenylsulfonyl)glycine Methyl Ester 98%
N-(Phenylsulfonyl)glycine Methyl Ester

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄S
MDL Number: MFCD00545497
CAS Number: 69398-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR