2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(pent-4-en-1-yl)hept-6-enoic acid 95%

Supplier: Ambeed
Warning

A477880-100MG A477880-250MG A477880-1G A477880-5G
76829-968EA 54.11 USD
76829-968 76829-970 77551-454 77551-456
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(pent-4-en-1-yl)hept-6-enoic acid 95%
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(pent-4-en-1-yl)hept-6-enoic acid

Precaución: for Research Use Only

Formula: C₂₇H₃₁NO₄
MW: 433.54 g/mol
MDL Number: MFCD12925762
CAS Number: 1068435-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR