N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine 97%

Supplier: Ambeed
Warning

Synonyms: N1-(4-Fluorobenzyl)-3-nitrobenzene-1,4-diamine, N1-(4-Fluorobenzyl)-3-nitro-1,4-phenylenediamine

A476910-1G A476910-250MG A476910-5G
76829-956EA 29.71 USD
76829-956 76829-958 76829-960
N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine 97%
N-(4-Fluorobenzyl)-3-nitro-p-phenylenediamine

Precaución: for Research Use Only

Formula: C₁₃H₁₂FN₃O₂
MW: 261.256 g/mol
MDL Number: MFCD12828054
CAS Number: 150812-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR