(E)-3-(2,4-Dimethoxybenzylidene)-3,4,5,6-tetrahydro-2,3'-bipyridine dihydrochloride 98+%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: GTS-21 2HCl

A475880-10MG A475880-1MG A475880-25MG A475880-50MG A475880-5MG A475880-250MG
76829-938EA 68 USD
76829-938 76829-940 76829-942 76829-944 76829-946 77684-270
(E)-3-(2,4-Dimethoxybenzylidene)-3,4,5,6-tetrahydro-2,3'-bipyridine dihydrochloride 98+%, Ambeed.Inc
(E)-3-(2,4-Dimethoxybenzylidene)-3,4,5,6-tetrahydro-2,3'-bipyridine dihydrochloride

Precaución: For research use only.

Formula: C₁₉H₂₂Cl₂N₂O₂
MW: 381.29 g/mol
MDL Number: MFCD18910849
CAS Number: 156223-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR