(2-Isopropoxy-4-(trifluoromethyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A475211-1G A475211-250MG A475211-5G
76829-932EA 46 USD
76829-932 76829-934 76829-936
(2-Isopropoxy-4-(trifluoromethyl)phenyl)boronic acid 98%
(2-Isopropoxy-4-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BF₃O₃
MDL Number: MFCD04972374
CAS Number: 1072952-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR