2-Methoxypyridine-3,4-diamine 95%

Supplier: Ambeed
Warning

A472650-100MG A472650-1G
76829-924EA 51.99 USD
76829-924 76829-926
2-Methoxypyridine-3,4-diamine 95%
2-Methoxypyridine-3,4-diamine

Precaución: for Research Use Only

Formula: C₆H₉N₃O
MW: 139.155 g/mol
MDL Number: MFCD10574697
CAS Number: 33631-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR