6-Chloro-5-methyl-3-nitropyridin-2-amine 97%

Supplier: Ambeed
Warning

A470140-100MG A470140-1G A470140-250MG A470140-5G
76829-914EA 78.51 USD
76829-914 76829-916 76829-918 77684-266
6-Chloro-5-methyl-3-nitropyridin-2-amine 97%
6-Chloro-5-methyl-3-nitropyridin-2-amine

Precaución: For research use only.

Formula: C₆H₆ClN₃O₂
MW: 187.58 g/mol
MDL Number: MFCD17014970
CAS Number: 202217-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR