1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

Synonyms: 2'-O-MOE-5-Me-rU

A469600-1G A469600-5G A469600-250MG
76829-910EA 55 USD
76829-910 76829-912 77886-220
1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione 95%
1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₇
MW: 316.31 g/mol
MDL Number: MFCD02682964
CAS Number: 163759-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR