1-Methyl-1H-indazole-4-boronic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (1-Methyl-1H-indazol-4-yl)boronic acid hydrochloride

A469162-100MG A469162-1G A469162-250MG
76829-900EA 36.08 USD
76829-900 76829-902 76829-904
1-Methyl-1H-indazole-4-boronic acid hydrochloride 95%
1-Methyl-1H-indazole-4-boronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀BClN₂O₂
MW: 212.4400 g/mol
MDL Number: MFCD13176537
CAS Number: 1257527-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR