6-(Dimethylamino)-2-naphthoic acid 97%

Supplier: Ambeed
Warning

A467657-1G A467657-250MG A467657-100MG
76829-892EA 255 USD
76829-892 76829-894 77886-215
6-(Dimethylamino)-2-naphthoic acid 97%
6-(Dimethylamino)-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
MDL Number: MFCD23135704
CAS Number: 5043-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR