2,5-Dichloro-4-methoxypyrimidine 97%

Supplier: Ambeed
Warning

A466210-1G A466210-250MG A466210-5G
76829-878EA 41.38 USD
76829-878 76829-880 76829-882
2,5-Dichloro-4-methoxypyrimidine 97%
2,5-Dichloro-4-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂O
MDL Number: MFCD11858598
CAS Number: 5750-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR