3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Chloro-2-fluoropyridne-5-boronic acid pinacol ester

A465548-100MG A465548-1G A465548-250MG
76829-872EA 34 USD
76829-872 76829-874 76829-876
3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester 98%
3-Chloro-2-fluoropyridine-5-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BClFNO₂
MW: 257.497 g/mol
MDL Number: MFCD16995268
CAS Number: 1220219-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR