3-Bromo-N,N-dimethylbenzamide 98%

Supplier: Ambeed
Warning

A463317-1G A463317-250MG A463317-25G A463317-5G
76829-852EA 42.44 USD
76829-852 76829-854 76829-856 76829-858
3-Bromo-N,N-dimethylbenzamide 98%
3-Bromo-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MW: 228.0858 g/mol
MDL Number: MFCD01215179
CAS Number: 24167-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR