(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 98+%

Supplier: Ambeed
Warning

Synonyms: (R)-4-Boc-morpholine-3-carboxylic acid

A461760-1G A461760-25G A461760-5G A461760-100G
76829-840EA 35 USD
76829-840 76829-842 76829-844 77886-206
(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 98+%
(R)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₅
MDL Number: MFCD04114909
CAS Number: 869681-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR