4-(Trifluoromethyl)-N,N-dimethylbenzamide 98%

Supplier: Ambeed
Warning

A461613-1G A461613-250MG A461613-25G
76829-834EA 37 USD
76829-834 76829-836 76829-838
4-(Trifluoromethyl)-N,N-dimethylbenzamide 98%
4-(Trifluoromethyl)-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃NO
MDL Number: MFCD16618942
CAS Number: 25771-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR