N-Boc-4-chloroaniline 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-chlorophenyl)carbamate

A460843-100G A460843-25G A460843-500G A460843-5G
76829-818EA 115.64 USD
76829-818 76829-820 76829-822 76829-824
N-Boc-4-chloroaniline 98%
N-Boc-4-chloroaniline

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO₂
MW: 227.688 g/mol
MDL Number: MFCD00816772
CAS Number: 18437-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR