1-Fluoro-3-(methoxymethoxy)benzene 95%

Supplier: Ambeed
Warning

A460084-1G A460084-250MG A460084-25G A460084-5G
76829-810EA 13 USD
76829-810 76829-812 76829-814 76829-816
1-Fluoro-3-(methoxymethoxy)benzene 95%
1-Fluoro-3-(methoxymethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MDL Number: MFCD09909406
CAS Number: 126940-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR