(S)-3,3'-Bis(3,5-dimethylphenyl)-1,1'-bi-2-naphthalene]-2,2'-diol ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (S)-3,3'-Bis(3,5-dimethylphenyl)-[1,1'-binaphthalene]-2,2'-diol

A459203-1G A459203-250MG
76829-802EA 371.32 USD
76829-802 76829-804
(S)-3,3'-Bis(3,5-dimethylphenyl)-1,1'-bi-2-naphthalene]-2,2'-diol ≥98%, ee 99%
(S)-3,3'-Bis(3,5-dimethylphenyl)-1,1'-bi-2-naphthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₃₆H₃₀O₂
MW: 494.622 g/mol
MDL Number: MFCD11617327
CAS Number: 435327-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR