2-Fluoro-5-isopropylphenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-5-isopropylphenyl)boronic acid

A458300-1G A458300-250MG
76829-796EA 297.06 USD
76829-796 76829-798
2-Fluoro-5-isopropylphenylboronic acid 96%
2-Fluoro-5-isopropylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BFO₂
MW: 182 g/mol
MDL Number: MFCD17015760
CAS Number: 1256354-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR