4,5-Dibromopyridin-2-amine 98%

Supplier: Ambeed
Warning

A457065-100MG A457065-1G A457065-250MG A457065-5G
76829-768EA 15.92 USD
76829-768 76829-770 76829-772 76829-774
4,5-Dibromopyridin-2-amine 98%
4,5-Dibromopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂
MW: 251.91 g/mol
MDL Number: MFCD16556235
CAS Number: 856848-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR