2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Pyrrolidin-1-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A455814-1G A455814-250MG A455814-5G
76829-756EA 67.9 USD
76829-756 76829-758 76829-760
2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester 98%
2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₃BN₂O₂
MW: 274.17 g/mol
MDL Number: MFCD06798280
CAS Number: 1073354-41-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR