Methyl 2'-cyano-[1,1'-biphenyl]-3-carboxylate 97%

Supplier: Ambeed
Warning

A453437-1G A453437-250MG
76829-744EA 63.66 USD
76829-744 76829-746
Methyl 2'-cyano-[1,1'-biphenyl]-3-carboxylate 97%
Methyl 2'-cyano-[1,1'-biphenyl]-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₁NO₂
MW: 237.25 g/mol
MDL Number: MFCD05980890
CAS Number: 168618-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR