2,6-Diisopropylphenyl isocyanate 97%

Supplier: Ambeed
Danger

A453142-25G A453142-5G A453142-1G
76829-738EA 166 USD
76829-738 76829-740 77886-197
2,6-Diisopropylphenyl isocyanate 97%
2,6-Diisopropylphenyl isocyanate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO
MDL Number: MFCD00008882
CAS Number: 28178-42-9
UN: 2206
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR