Methyl-2-(3-oxetanylidene)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(oxetan-3-ylidene)acetate

A452911-1G A452911-250MG
76829-734EA 60.48 USD
76829-734 76829-736
Methyl-2-(3-oxetanylidene)acetate 98%
Methyl-2-(3-oxetanylidene)acetate

Precaución: for Research Use Only

Formula: C₆H₈O₃
MW: 128.13 g/mol
MDL Number: MFCD16140399
CAS Number: 1105665-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR