(1R)-1-(2-Fluorophenyl)ethan-1-ol 96%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Fluoro-α-methylbenzyl alcohol, (R)-1-(2-Fluorophenyl)ethanol

A451644-1G A451644-250MG A451644-5G
76829-718EA 29 USD
76829-718 76829-720 76829-722
(1R)-1-(2-Fluorophenyl)ethan-1-ol 96%
(1R)-1-(2-Fluorophenyl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₈H₉FO
MDL Number: MFCD03092991
CAS Number: 162427-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR