Oxazolo[5,4-b]pyridine-2(1H)-thione 96%

Supplier: Ambeed
Warning

A451207-1G A451207-250MG A451207-5G
76829-712EA 162.32 USD
76829-712 76829-714 76829-716
Oxazolo[5,4-b]pyridine-2(1H)-thione 96%
Oxazolo[5,4-b]pyridine-2(1H)-thione

Precaución: for Research Use Only

Formula: C₆H₄N₂OS
MW: 152.174 g/mol
MDL Number: MFCD21908350
CAS Number: 169205-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR