3-Chloro-5-isopropoxyphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Chloro-5-isopropoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A450348-1G A450348-250MG A450348-5G
76829-704EA 200.52 USD
76829-704 76829-706 76829-708
3-Chloro-5-isopropoxyphenylboronic acid pinacol ester 98%
3-Chloro-5-isopropoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₂BClO₃
MW: 296.6 g/mol
MDL Number: MFCD12546618
CAS Number: 1218789-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR