1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole

A449153-1G A449153-25G A449153-5G
76829-696EA 23.34 USD
76829-696 76829-698 76829-700
1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole 98%
1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrN₂
MDL Number: MFCD11845717
CAS Number: 294877-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR