6-Chloro-4-trifluoromethylpyridin-2-ylamine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-chloro-4-(trifluoromethyl)pyridine, 6-Chloro-4-(trifluoromethyl)pyridin-2-amine

A448775-100MG A448775-250MG
76829-692EA 45.62 USD
76829-692 76829-694
6-Chloro-4-trifluoromethylpyridin-2-ylamine 95%
6-Chloro-4-trifluoromethylpyridin-2-ylamine

Precaución: for Research Use Only

Formula: C₆H₄ClF₃N₂
MW: 196.56 g/mol
MDL Number: MFCD00128898
CAS Number: 34486-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR