(R)-(+)-3-Bromo-α-methylbenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Bromo-α-methylbenzyl alcohol, (R)-3-Bromo-alpha-methylbenzyl Alcohol

A443351-1G A443351-250MG A443351-5G
76829-666EA 14 USD
76829-666 76829-668 77594-388
(R)-(+)-3-Bromo-α-methylbenzyl alcohol 95%
(R)-(+)-3-Bromo-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MDL Number: MFCD06658929
CAS Number: 134615-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR