1,3-Dibromo-5-phenoxybenzene 98%

Supplier: Ambeed
Warning

A443155-10G A443155-1G A443155-250MG A443155-5G
76829-658EA 318 USD
76829-658 76829-660 76829-662 76829-664
1,3-Dibromo-5-phenoxybenzene 98%
1,3-Dibromo-5-phenoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₈Br₂O
MDL Number: MFCD17018983
CAS Number: 46438-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR