6-Bromo-4-chloroquinoline-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A442149-1G A442149-250MG
76829-646EA 40 USD
76829-646 76829-648
6-Bromo-4-chloroquinoline-2-carboxylic acid 95%
6-Bromo-4-chloroquinoline-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₅BrClNO₂
MDL Number: MFCD06810525
CAS Number: 887589-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR