Methyl 3-chloro-2-fluorobenzoate 98%

Supplier: Ambeed
Warning

A441261-1G A441261-5G A441261-25G
76829-632EA 17 USD
76829-632 76829-634 77886-173
Methyl 3-chloro-2-fluorobenzoate 98%
Methyl 3-chloro-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MDL Number: MFCD06203787
CAS Number: 773874-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR