4-(4-Aminophenyl)-3-methyl-4-oxobutanoic acid hydrochloride 95+%

Supplier: Ambeed
Warning

A441129-25G A441129-1G
76829-630EA 82.76 USD
76829-630 77684-213
4-(4-Aminophenyl)-3-methyl-4-oxobutanoic acid hydrochloride 95+%
4-(4-Aminophenyl)-3-methyl-4-oxobutanoic acid hydrochloride

Precaución: For research use only.

Formula: C₁₁H₁₄ClNO₃
MW: 243.68 g/mol
MDL Number: MFCD23135502
CAS Number: 120757-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR