tert-Butyl 3-(aminomethyl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Danger

A440263-250MG A440263-100MG A440263-1G
76829-628EA 85 USD
76829-628 77886-165 77886-166
tert-Butyl 3-(aminomethyl)piperazine-1-carboxylate 95%
tert-Butyl 3-(aminomethyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₂₁N₃O₂
MW: 215.29 g/mol
MDL Number: MFCD22056449
CAS Number: 1376099-80-7
UN: 2735
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR