(4-Bromophenyl)acetamide 96%

Supplier: Ambeed
Warning

A439519-1G A439519-250MG A439519-5G
76829-614EA 35 USD
76829-614 76829-616 76829-618
(4-Bromophenyl)acetamide 96%
(4-Bromophenyl)acetamide
Formula: C₈H₈BrNO
MDL Number: MFCD00126843
CAS Number: 74860-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR