(5-(Methoxycarbonyl)-1H-pyrrol-2-yl)boronic acid 98%

Supplier: Ambeed
Warning

A435574-1G A435574-250MG A435574-5G
76829-588EA 131 USD
76829-588 76829-590 76829-592
(5-(Methoxycarbonyl)-1H-pyrrol-2-yl)boronic acid 98%
(5-(Methoxycarbonyl)-1H-pyrrol-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₄
MW: 168.94 g/mol
MDL Number: MFCD15143454
CAS Number: 1217500-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR