1-Hexanesulphonic acid sodium salt monohydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium 1-hexanesulphonate monohydrate
A435405-5G
A435405-100G
A435405-25G
76829-586EA
17
USD
76829-586
77551-304
77886-155
1-Hexanesulphonic acid sodium salt monohydrate 98%
1-Hexanesulphonic acid sodium salt monohydrate
Precaución: for Research Use Only
Formula:
C₆H₁₃NaO₃S·1H₂O MW: 206.24 g/mol |
MDL Number:
MFCD00149549 CAS Number: 207300-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...