Ethyl 3-hydroxycyclohexanecarboxylate 98%

Supplier: Ambeed
Warning

A433847-1G A433847-250MG A433847-5G
76829-568EA 62.6 USD
76829-568 76829-570 76829-572
Ethyl 3-hydroxycyclohexanecarboxylate 98%
Ethyl 3-hydroxycyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₆O₃
MW: 172.222 g/mol
MDL Number: MFCD00205586
CAS Number: 94160-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR