6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7(8H)-one 98%

Supplier: Ambeed
Warning

A432404-100G A432404-10G A432404-1G A432404-25G A432404-5G
76829-550EA 212.18 USD
76829-550 76829-552 76829-554 76829-556 76829-558
6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7(8H)-one 98%
6-Bromo-2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7(8H)-one

Precaución: for Research Use Only

Formula: C₁₃H₁₃BrClN₃O
MW: 342.61 g/mol
MDL Number: MFCD13181208
CAS Number: 1016636-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR