2-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, 2-(2-Methoxy-2-oxoethyl)benzeneboronic acid, pinacol ester

A430862-100MG A430862-1G A430862-250MG A430862-5G A430862-25G
76829-542EA 11 USD
76829-542 76829-544 76829-546 76829-548 77886-142
2-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester 98%
2-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₄
MDL Number: MFCD12913983
CAS Number: 956229-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR