2-Bromo-5-fluoro-4-nitropyridine 1-oxide 95%

Supplier: Ambeed
Warning

A430689-100MG A430689-250MG
76829-534EA 28 USD
76829-534 76829-536
2-Bromo-5-fluoro-4-nitropyridine 1-oxide 95%
2-Bromo-5-fluoro-4-nitropyridine 1-oxide

Precaución: for Research Use Only

Formula: C₅H₂BrFN₂O₃
MDL Number: MFCD09261237
CAS Number: 935534-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR